Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197068.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197068.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197068.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197068.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197068.hg.1 |
TC1200012403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197067.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197067.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197067.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197067.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197067.hg.1 |
TC1200012403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200197066.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197066.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197066.hg.1 |
ENST00000540745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197066.hg.1 |
NR_104647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197066.hg.1 |
TC1200012403.hg.1 |
Clariom D TC |