Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182019.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182019.hg.1 |
TC1600010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600182019.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010637.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182016.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182016.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182016.hg.1 |
TC1600010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600182016.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182016.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182011.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182011.hg.1 |
TC1600010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600182011.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182014.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182014.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182014.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182014.hg.1 |
TC1600010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600182014.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182014.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182014.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182013.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182013.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182013.hg.1 |
TC1600010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600182013.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182013.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182017.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182017.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182017.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182017.hg.1 |
TC1600010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600182017.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182017.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182017.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182018.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182018.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182018.hg.1 |
TC1600010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600182018.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182018.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182015.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182015.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182015.hg.1 |
NR_104656.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182015.hg.1 |
TC1600010637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600182015.hg.1 |
ENST00000613438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182015.hg.1 |
ENST00000621378.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182015.hg.1 |
NR_104656.1 |
Refseq |