Assay Details
Gene Symbol: LINC01229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104660.1 | 3 - 4 | 548 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MAF transcriptional regulator RNA | MAFTRR |
linc-MAF-4, lincMAF4 |
-27.9kb | Anti-sense | Chr. 16 - 79721312 - 79770532 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC102724084 | LOC102724084 | 354.6kb | Anti-sense | Chr. 16 - 80155958 - 80563135 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | TC1600011435.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191603.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | TC1600011435.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191602.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191610.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191610.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191610.hg.1 | TC1600011435.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600191610.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191610.hg.1 | NR_104660.1 | Refseq |
