Assay Details
Gene Symbol: LOC101927488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105039.1 | 2 - 3 | 349 | 140 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105369178 | LOC105369178 | Sense | Chr. 5 - 125980474 - 126368825 on NCBI Build GRCh38 | ||
Downstream | GRAM domain containing 3 | GRAMD3 | NS3TP2 | 80.2kb | Anti-sense | Chr. 5 - 126360096 - 126494377 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
