Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158473.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158473.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158473.hg.1 |
TC0500008534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500158473.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158473.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
ENST00000507428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008534.hg.1 |
NR_105042.1 |
Refseq |