Assay Details
Gene Symbol: LOC101927835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105064.1 | 3 - 4 | 307 | 88 | ||
GenBank mRNA | AW182569.1 | - | 1 - 2 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methionine adenosyltransferase 2B | MAT2B |
MAT-II, MATIIbeta, Nbla02999, SDR23E1, TGR |
-929.1kb | Anti-sense | Chr. 5 - 163503064 - 163519353 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC102546299 | LOC102546299 | 20.9kb | Anti-sense | Chr. 5 - 164470279 - 164542984 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
