Assay Details
Gene Symbol: MIR6134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_106750.1 | 1 | 54 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DDB1 and CUL4 associated factor 8 like 1 | DCAF8L1 | WDR42B | -514.1kb | Sense | Chr. X - 27977993 - 27981449 on NCBI Build GRCh38 |
Downstream | interleukin 1 receptor accessory protein like 1 | IL1RAPL1 |
IL1R8, IL1RAPL, MRX10, MRX21, MRX34, OPHN4, TIGIRR-2 |
91.9kb | Anti-sense | Chr. X - 28587564 - 29956350 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009306.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
