Assay Details
Gene Symbol: MIR6865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_106925.1 | 1 | 37 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6864 | MIR6864 | hsa-mir-6864 | -315.0 | Sense | Chr. 17 - 4969702 - 4969771 on NCBI Build GRCh38 |
Downstream | translation initiation factor IF-2 | LOC101927979 | 17.5kb | Anti-sense | Chr. 17 - 4986991 - 4990556 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700009543.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
