Assay Details
Gene Symbol: LINC00845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_108052.1 | 1 - 2 | 70 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Rho related BTB domain containing 1 | RHOBTB1 | -14.8kb | Anti-sense | Chr. 10 - 60869438 - 61001486 on NCBI Build GRCh38 | |
Downstream | transmembrane protein 26 | TMEM26 | 380.2kb | Anti-sense | Chr. 10 - 61406642 - 61453450 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
