Assay Details
Gene Symbol: LINC00339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109759.1 | 1 - 2 | 140 | 85 | ||
GenBank mRNA | BX509008.1 | - | 1 - 2 | 141 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928043 | LOC101928043 | Anti-sense | Chr. 1 - 22023994 - 22026048 on NCBI Build GRCh38 | ||
Downstream | cell division cycle 42 | CDC42 |
CDC42Hs, G25K, TKS |
21.4kb | Sense | Chr. 1 - 22052627 - 22092943 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000452322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000452322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109762.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000416769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000420503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000434233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000452322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000452322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023918.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_023919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109759.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | NR_109761.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007267.hg.1 | ENST00000404210.2 | Ensembl |
