Assay Details
Gene Symbol: LINC00339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109761.1 | 1 - 2 | 137 | 97 | ||
GenBank mRNA | BG432520.1 | - | 1 - 2 | 131 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928043 | LOC101928043 | Anti-sense | Chr. 1 - 22023994 - 22026048 on NCBI Build GRCh38 | ||
Downstream | cell division cycle 42 | CDC42 |
CDC42Hs, G25K, TKS |
25.1kb | Sense | Chr. 1 - 22052627 - 22092943 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
