Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007886.hg.1 |
ENST00000446201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007886.hg.1 |
ENST00000446201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007886.hg.1 |
ENST00000446201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153166.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153166.hg.1 |
TC0900007886.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153166.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153169.hg.1 |
ENST00000446201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153169.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153169.hg.1 |
TC0900007886.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153169.hg.1 |
ENST00000446201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153169.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007886.hg.1 |
ENST00000446201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007886.hg.1 |
ENST00000446201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007886.hg.1 |
ENST00000446201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007886.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153167.hg.1 |
NR_109791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153167.hg.1 |
TC0900007886.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153167.hg.1 |
NR_109791.1 |
Refseq |