Assay Details
Gene Symbol: LINC01470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109877.1 | 4 - 5 | 364 | 114 | ||
GenBank mRNA | AK123816.1 | - | 4 - 5 | 364 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neuromedin U receptor 2 | NMUR2 |
FM-4, FM4, NMU-R2, NMU2R, TGR-1, TGR1 |
-213.7kb | Sense | Chr. 5 - 152391541 - 152405279 on NCBI Build GRCh38 |
Downstream | glutamate ionotropic receptor AMPA type subunit 1 | GRIA1 |
GLUH1, GLUR1, GLURA, GluA1, HBGR1 |
804.9kb | Anti-sense | Chr. 5 - 153489615 - 153813873 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187489.hg.1 | TC0500012533.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187487.hg.1 | TC0500012533.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500187485.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187485.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187485.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187485.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187485.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187485.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187485.hg.1 | TC0500012533.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187494.hg.1 | TC0500012533.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500187500.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187500.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187500.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187500.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187500.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187500.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187500.hg.1 | TC0500012533.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000503048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000511419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | ENST00000584829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012533.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | ENST00000506723.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187486.hg.1 | TC0500012533.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500187491.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187491.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187491.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187491.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187491.hg.1 | ENST00000522300.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187491.hg.1 | NR_109877.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187491.hg.1 | TC0500012533.hg.1 | Clariom D TC |
