Assay Details
Gene Symbol: LOC101929625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109880.1 | 1 - 2 | 221 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101929608 | LOC101929608 | Anti-sense | Chr. 20 - 21610787 - 21703585 on NCBI Build GRCh38 | ||
Downstream | paired box 1 | PAX1 |
HUP48, OFC2 |
93.0kb | Sense | Chr. 20 - 21705659 - 21718486 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147296.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147296.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147296.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147296.hg.1 | TC2000006909.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147296.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147296.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147296.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147299.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147299.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147299.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147299.hg.1 | TC2000006909.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147299.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147299.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147299.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147300.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147300.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147300.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147300.hg.1 | TC2000006909.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147300.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147300.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147300.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147294.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147294.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147294.hg.1 | TC2000006909.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147294.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147294.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
