Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009919.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169806.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169806.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169806.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169806.hg.1 |
TC2000009919.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169806.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169806.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169806.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169805.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169805.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169805.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169805.hg.1 |
TC2000009919.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169805.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169805.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169805.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009919.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169801.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169801.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169801.hg.1 |
TC2000009919.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169801.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169801.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
NR_109937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
TC2000009919.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
ENST00000432633.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
ENST00000445606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
NR_109936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169804.hg.1 |
NR_109937.1 |
Refseq |