Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
ENST00000509228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
ENST00000509228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010184.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
ENST00000509228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
ENST00000509228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010184.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171280.hg.1 |
ENST00000509228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171280.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171280.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171280.hg.1 |
TC0500010184.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500171280.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171280.hg.1 |
ENST00000509228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171280.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171278.hg.1 |
ENST00000510456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500171278.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171278.hg.1 |
TC0500010184.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500171278.hg.1 |
NR_109945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500171278.hg.1 |
ENST00000510456.1 |
Ensembl |