Assay Details
Gene Symbol: LOC101927881 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110278.1 | 2 - 3 | 220 | 81 | ||
GenBank mRNA | AK311291.1 | - | 2 - 3 | 215 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | heparan sulfate 6-O-sulfotransferase 1 | HS6ST1 |
HH15, HS6ST |
-547.1kb | Anti-sense | Chr. 2 - 128265480 - 128318597 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC151121 | LOC151121 | 373.4kb | Anti-sense | Chr. 2 - 129242173 - 129273891 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
