Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000440545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000440545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000440545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000440545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228767.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228767.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228767.hg.1 |
TC0200016765.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228767.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228767.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228764.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228764.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228764.hg.1 |
TC0200016765.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228764.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228764.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228771.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228771.hg.1 |
TC0200016765.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228771.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000417096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000432413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000440545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000440545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000440545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000440545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
ENST00000448786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016765.hg.1 |
NR_110283.1 |
Refseq |