Assay Details
Gene Symbol: LOC101927865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110283.1 | 2 - 3 | 193 | 70 | ||
GenBank mRNA | HY006998.1 | - | 2 - 3 | 194 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Kruppel like factor 7 | KLF7 | UKLF | -64.0kb | Sense | Chr. 2 - 207074137 - 207173851 on NCBI Build GRCh38 |
Downstream | microRNA 1302-4 | MIR1302-4 |
MIRN1302-4, hsa-mir-1302-4 |
28.9kb | Sense | Chr. 2 - 207269275 - 207269424 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
