Assay Details
Gene Symbol: LOC101929733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110575.1 | 1 - 2 | 182 | 56 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein disulfide isomerase family A member 6 | PDIA6 |
ERP5, P5, TXNDC7 |
-40.3kb | Anti-sense | Chr. 2 - 10783391 - 10837977 on NCBI Build GRCh38 |
Downstream | potassium voltage-gated channel modifier subfamily F member 1 | KCNF1 |
IK8, KCNF, KV5.1, kH1 |
26.8kb | Sense | Chr. 2 - 10911937 - 10914225 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
