Assay Details
Gene Symbol: LOC101929715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110597.1 | 1 - 2 | 138 | 104 | ||
GenBank mRNA | AW303360.1 | - | 3 - 4 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ornithine decarboxylase 1 | ODC1 | ODC | -1.2kb | Anti-sense | Chr. 2 - 10440371 - 10448554 on NCBI Build GRCh38 |
Downstream | nucleolar protein 10 | NOL10 | PQBP5 | 120.5kb | Anti-sense | Chr. 2 - 10570766 - 10689991 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000404616.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006692.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | TC0200006692.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145857.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | TC0200006692.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145859.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | NR_110597.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | TC0200006692.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | ENST00000547349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | ENST00000553181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200145860.hg.1 | NR_110597.1 | Refseq |
