Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145874.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145874.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145874.hg.1 |
TC0200006692.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200145874.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145874.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145875.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145875.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145875.hg.1 |
TC0200006692.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200145875.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145875.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000404616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000547349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
ENST00000553181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006692.hg.1 |
NR_110597.1 |
Refseq |