Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176031.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176031.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176031.hg.1 |
TC0200010716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176031.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176031.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176026.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176026.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176026.hg.1 |
TC0200010716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176026.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176026.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010716.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176029.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176029.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176029.hg.1 |
TC0200010716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176029.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176029.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176032.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176032.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176032.hg.1 |
TC0200010716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176032.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176032.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176033.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176033.hg.1 |
NR_110607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176033.hg.1 |
TC0200010716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176033.hg.1 |
ENST00000447835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176033.hg.1 |
NR_110607.1 |
Refseq |