Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009277.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009277.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009277.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009277.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009277.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009277.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009277.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009277.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173959.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173959.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173959.hg.1 |
TC1900009277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900173959.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173959.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009277.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009277.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009277.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009277.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009277.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009277.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009277.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009277.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173958.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173958.hg.1 |
NR_110670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173958.hg.1 |
TC1900009277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900173958.hg.1 |
ENST00000587587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173958.hg.1 |
NR_110670.1 |
Refseq |