Assay Details
Gene Symbol: LINC01351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110716.1 | 2 - 3 | 290 | 94 | ||
GenBank mRNA | CR936711.1 | - | 2 - 3 | 291 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | BMP/retinoic acid inducible neural specific 3 | BRINP3 |
DBCCR1L, DBCCR1L1, FAM5C |
-678.0 | Anti-sense | Chr. 1 - 190097662 - 190477997 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC440704 | LOC440704 | 143.5kb | Sense | Chr. 1 - 190624890 - 190801658 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000417409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000417409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000417409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240083.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240083.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240083.hg.1 | TC0100018326.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100240083.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240083.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | TC0100018326.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240076.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | TC0100018326.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240079.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | TC0100018326.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240080.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000417409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000417409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000417409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018326.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | ENST00000417409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | TC0100018326.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | ENST00000417409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240077.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | TC0100018326.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | ENST00000424735.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | ENST00000436905.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240078.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240082.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240082.hg.1 | NR_110717.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240082.hg.1 | TC0100018326.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100240082.hg.1 | NR_110716.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240082.hg.1 | NR_110717.1 | Refseq |
