Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010695.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010695.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010695.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010695.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010695.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010695.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010695.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010695.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600172660.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600172660.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600172660.hg.1 |
TC0600010695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600172660.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600172660.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600172662.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600172662.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600172662.hg.1 |
TC0600010695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600172662.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600172662.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600172661.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600172661.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600172661.hg.1 |
TC0600010695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600172661.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600172661.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010695.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010695.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010695.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010695.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010695.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010695.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010695.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010695.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600172663.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600172663.hg.1 |
NR_110842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600172663.hg.1 |
TC0600010695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600172663.hg.1 |
ENST00000602500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600172663.hg.1 |
NR_110842.1 |
Refseq |