Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197647.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197647.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197647.hg.1 |
TC1700011955.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197647.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197647.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
ENST00000576738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011955.hg.1 |
NR_110850.1 |
Refseq |