Assay Details
Gene Symbol: LOC101927839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110888.1 | 1 - 2 | 52 | 89 | ||
GenBank mRNA | BI461260.1 | - | 1 - 2 | 59 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
SMG6, nonsense mediated mRNA decay factor |
SMG6 |
C17orf31, EST1A, SMG-6, hSMG5/7a |
Anti-sense | Chr. 17 - 2059839 - 2303836 on NCBI Build GRCh38 | |
Downstream | serine racemase | SRR |
ILV1, ISO1 |
69.8kb | Sense | Chr. 17 - 2303371 - 2325267 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
