Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011334.hg.1 |
NR_110949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188939.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188939.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188939.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188939.hg.1 |
TC1600011334.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600188939.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188939.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188939.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
ENST00000572086.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011334.hg.1 |
NR_110949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188941.hg.1 |
NR_110947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188941.hg.1 |
TC1600011334.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600188941.hg.1 |
NR_110947.1 |
Refseq |