Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024597.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024597.hg.1 |
ENST00000415700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024597.hg.1 |
NR_033880.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024597.hg.1 |
TC02001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000415700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000415700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000415700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024596.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024596.hg.1 |
NR_033880.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024596.hg.1 |
TC02001502.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000414556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000415700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000415700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000415700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011476.hg.1 |
ENST00000621134.4 |
Ensembl |