Assay Details
Gene Symbol: GRTP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120385.1 | 1 - 2 | 296 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | growth hormone regulated TBC protein 1 | GRTP1 | TBC1D6 | Anti-sense | Chr. 13 - 113324164 - 113364158 on NCBI Build GRCh38 | |
Downstream | collagen alpha-1(II) chain-like | LOC101928841 | 37.7kb | Anti-sense | Chr. 13 - 113399607 - 113420770 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
