Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000412119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000412119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000412119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000412119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154217.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154217.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154217.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154217.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154217.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154217.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154217.hg.1 |
TC1300008054.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154216.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154216.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154216.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154216.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154216.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154216.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154216.hg.1 |
TC1300008054.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300154218.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154218.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154218.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154218.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154218.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154218.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154218.hg.1 |
TC1300008054.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000412119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000412119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000412119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000412119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
ENST00000440860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_046992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008054.hg.1 |
NR_120401.1 |
Refseq |