Assay Details
Gene Symbol: LOC102723885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120517.1 | 1 - 2 | 98 | 102 | ||
GenBank mRNA | DB043921.1 | - | 1 - 2 | 98 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ADAM metallopeptidase domain 22 | ADAM22 |
ADAM 22, MDC2 |
-13.8kb | Sense | Chr. 7 - 87934143 - 88202889 on NCBI Build GRCh38 |
Downstream | STEAP4 metalloreductase | STEAP4 |
STAMP2, TIARP, TNFAIP9 |
57.2kb | Anti-sense | Chr. 7 - 88276429 - 88306913 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
