Assay Details
Gene Symbol: LOC101928940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120567.1 | 1 - 2 | 50 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nicotinamide N-methyltransferase | NNMT | -66.2kb | Anti-sense | Chr. 11 - 114295813 - 114312516 on NCBI Build GRCh38 | |
Downstream | chromosome 11 open reading frame 71 | C11orf71 | URLC7 | 11.4kb | Sense | Chr. 11 - 114391448 - 114400550 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
