Assay Details
Gene Symbol: LOC101928940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120567.1 | 2 - 3 | 298 | 97 | ||
GenBank mRNA | BC033908.1 | - | 2 - 3 | 303 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nicotinamide N-methyltransferase | NNMT | -48.1kb | Anti-sense | Chr. 11 - 114295813 - 114312516 on NCBI Build GRCh38 | |
Downstream | chromosome 11 open reading frame 71 | C11orf71 | URLC7 | 12.5kb | Sense | Chr. 11 - 114391448 - 114400550 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
