Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000432699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000432699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000432699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000611797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000611797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
ENST00000611797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000432699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000432699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000432699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000611797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000611797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
ENST00000611797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009213.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163197.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163197.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163197.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163197.hg.1 |
TC1000009213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000163197.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163197.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163197.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163198.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163198.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163198.hg.1 |
TC1000009213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000163198.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163198.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163196.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163196.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163196.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163196.hg.1 |
TC1000009213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000163196.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163196.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163196.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
ENST00000432699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
NR_120631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
TC1000009213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
ENST00000432699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
ENST00000448422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
NR_120630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163195.hg.1 |
NR_120631.1 |
Refseq |