Assay Details
Gene Symbol: LOC102723439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120673.1 | 2 - 3 | 262 | 187 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | adaptor related protein complex 3 mu 1 subunit | AP3M1 | -358.3kb | Sense | Chr. 10 - 74120257 - 74151085 on NCBI Build GRCh38 | |
Downstream | lysine acetyltransferase 6B | KAT6B |
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf |
296.9kb | Anti-sense | Chr. 10 - 74824927 - 75032623 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | TC1000011055.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175456.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | TC1000011055.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175452.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175445.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175445.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175445.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175445.hg.1 | TC1000011055.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000175445.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175445.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175445.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | TC1000011055.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175455.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011055.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | TC1000011055.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175451.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | TC1000011055.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000595410.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | ENST00000609392.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175454.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175448.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175448.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175448.hg.1 | NR_120673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000175448.hg.1 | TC1000011055.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000175448.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175448.hg.1 | ENST00000448214.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000175448.hg.1 | NR_120673.1 | Refseq |
