Assay Details
Gene Symbol: LINC01509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121581.1 | 2 - 3 | 278 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
RAD23 homolog B, nucleotide excision repair protein |
RAD23B |
HHR23B, HR23B, P58 |
-116.2kb | Anti-sense | Chr. 9 - 107283236 - 107332194 on NCBI Build GRCh38 |
Downstream | Kruppel like factor 4 | KLF4 |
EZF, GKLF |
29.9kb | Sense | Chr. 9 - 107484852 - 107489720 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
