Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166605.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166605.hg.1 |
TC0900009739.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900166605.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166609.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166609.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166609.hg.1 |
TC0900009739.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900166609.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166609.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166606.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166606.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166606.hg.1 |
TC0900009739.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900166606.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166606.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
ENST00000440888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009739.hg.1 |
NR_121602.1 |
Refseq |