Assay Details
Gene Symbol: LOC101929563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121602.1 | 6 - 7 | 962 | 114 | ||
GenBank mRNA | CR627240.1 | - | 6 - 7 | 950 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1239 | LINC01239 | -825.0kb | Anti-sense | Chr. 9 - 22646200 - 22824213 on NCBI Build GRCh38 | |
Downstream | ELAV like neuron-specific RNA binding protein 2 | ELAVL2 |
HEL-N1, HELN1, HUB |
39.0kb | Sense | Chr. 9 - 23690099 - 23850757 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
