Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222466.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222466.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222466.hg.1 |
TC0100016125.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100222466.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222466.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222465.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222465.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222465.hg.1 |
TC0100016125.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100222465.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222465.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016125.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222464.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222464.hg.1 |
NR_123725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222464.hg.1 |
TC0100016125.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100222464.hg.1 |
ENST00000418602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222464.hg.1 |
NR_123725.1 |
Refseq |