Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
ENST00000456532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
ENST00000456532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
ENST00000456532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
NR_125365.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
NR_125365.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011362.hg.1 |
NR_125365.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
ENST00000456532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
ENST00000456532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
ENST00000456532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
NR_125365.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
NR_125365.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011362.hg.1 |
NR_125365.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178649.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178649.hg.1 |
ENST00000456532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178649.hg.1 |
NR_125365.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178649.hg.1 |
ENST00000414772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178649.hg.1 |
ENST00000456532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178649.hg.1 |
NR_125365.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178649.hg.1 |
TC0X00011362.hg.1 |
Clariom D TC |