Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012704.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012704.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012704.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012704.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012704.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012704.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012704.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012704.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012704.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012704.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012704.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012704.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197680.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197680.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197680.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197680.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197680.hg.1 |
TC1100012704.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100197681.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197681.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197681.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197681.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197681.hg.1 |
TC1100012704.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100197679.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197679.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197679.hg.1 |
ENST00000532316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197679.hg.1 |
NR_125759.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197679.hg.1 |
TC1100012704.hg.1 |
Clariom D TC |