Assay Details
Gene Symbol: LOC102723729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125824.1 | 1 - 2 | 57 | 74 | ||
GenBank mRNA | BE326543.1 | - | 1 - 2 | 57 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glycerol-3-phosphate acyltransferase 4 | GPAT4 |
1-AGPAT 6, AGPAT6, LPAAT-zeta, LPAATZ, TSARG7 |
Anti-sense | Chr. 8 - 41578188 - 41625001 on NCBI Build GRCh38 | |
Downstream | NK6 homeobox 3 | NKX6-3 | NKX6.3 | 67.9kb | Sense | Chr. 8 - 41645177 - 41650818 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
