Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199042.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199042.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199042.hg.1 |
TC0600014146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600199042.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199042.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199043.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199043.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199043.hg.1 |
TC0600014146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600199043.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199043.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199045.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199045.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199045.hg.1 |
TC0600014146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600199045.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199045.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199044.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199044.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199044.hg.1 |
TC0600014146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600199044.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199044.hg.1 |
NR_125859.1 |
Refseq |