Assay Details
Gene Symbol: LOC101929762 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125931.1 | 2 - 3 | 295 | 92 | ||
GenBank mRNA | AK098126.1 | - | 3 - 4 | 479 | 92 | |
AK309988.1 | - | 2 - 3 | 295 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myozenin 2 | MYOZ2 |
C4orf5, CMH16, CS-1 |
-5.0kb | Anti-sense | Chr. 4 - 119135784 - 119187789 on NCBI Build GRCh38 |
Downstream | ubiquitin specific peptidase 53 | USP53 | 17.1kb | Anti-sense | Chr. 4 - 119212583 - 119295518 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
