Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009531.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009531.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009531.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009531.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009531.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009531.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009531.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009531.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168582.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168582.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168582.hg.1 |
TC0100009531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100168582.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168582.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168583.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168583.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168583.hg.1 |
TC0100009531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100168583.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168583.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168586.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168586.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168586.hg.1 |
TC0100009531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100168586.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168586.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009531.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009531.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009531.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009531.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009531.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009531.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009531.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009531.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168585.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168585.hg.1 |
NR_125969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168585.hg.1 |
TC0100009531.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100168585.hg.1 |
ENST00000438885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168585.hg.1 |
NR_125969.1 |
Refseq |