Assay Details
Gene Symbol: LOC101928977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125970.1 | 2 - 3 | 262 | 119 | ||
GenBank mRNA | DA297063.1 | - | 2 - 3 | 263 | 119 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928995 | LOC101928995 | Anti-sense | Chr. 1 - 115919376 - 115925908 on NCBI Build GRCh38 | ||
Downstream | solute carrier family 22 member 15 | SLC22A15 |
FLIPT1, PRO34686 |
16.9kb | Anti-sense | Chr. 1 - 115976498 - 116070054 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
