Assay Details
Gene Symbol: LPP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046623.1 | 1 - 2 | 289 | 70 | ||
GenBank mRNA | AW592678.1 | - | 1 - 2 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC401105 | FLJ42393 | AVPC1948 | -381.4kb | Anti-sense | Chr. 3 - 188178543 - 188180808 on NCBI Build GRCh38 |
Downstream | microRNA 28 | MIR28 |
MIRN28, hsa-mir-28, miR-28 |
120.1kb | Anti-sense | Chr. 3 - 188688781 - 188688866 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
