Assay Details
Gene Symbol: LINC00392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_047009.1 | 2 - 3 | 165 | 87 | ||
GenBank mRNA | AW592202.1 | - | 1 - 2 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Kruppel like factor 5 | KLF5 |
BTEB2, CKLF, IKLF |
-509.8kb | Sense | Chr. 13 - 73054976 - 73077542 on NCBI Build GRCh38 |
Downstream | Kruppel like factor 12 | KLF12 |
AP-2rep, AP2REP, HSPC122 |
98.1kb | Anti-sense | Chr. 13 - 73686012 - 74168295 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
